Learn More
Cayman Chemical DecnoIc AcId-d2 25mg
SDP
Supplier:  Cayman Chemical 2940925 MG
Encompass_Preferred
An internal standard for the quantification of decanoic acid by GC- or LC-MS
Catalog No. 50-299-0008