Learn More
Cayman Chemical DecnoIc AcId-d3 100mg
SDP
Catalog No. 502938289
Encompass_Preferred
An internal standard for the quantification of decanoic acid by GC- or LC-MS
Catalog No. 50-293-8289 Supplier Cayman Chemical Supplier No. 28516100 MG
An internal standard for the quantification of decanoic acid by GC- or LC-MS