Learn More
Cayman Chemical DecnoIc AcId-d3 10mg
SDP
Catalog No. 502940059
Encompass_Preferred
An internal standard for the quantification of decanoic acid by GC- or LC-MS
Catalog No. 50-294-0059 Supplier Cayman Chemical Supplier No. 2851610 MG
An internal standard for the quantification of decanoic acid by GC- or LC-MS