Learn More
Cayman Chemical Decanoic Acid-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 102611-15-4, 50 mg
SDP
Catalog No. 502937964
Encompass_Preferred
An internal standard for the quantification of decanoic acid by GC- or LC-MS
Catalog No. 50-293-7964 Supplier Cayman Chemical Supplier No. 2851650 MG
An internal standard for the quantification of decanoic acid by GC- or LC-MS