Learn More
Cayman Chemical Decanoyl-Coenzyme A (hydrate), Purity: ≥90%, 5 mg
SDP
Catalog No. 502938528
Encompass_Preferred
A thioester of decanoic acid and CoA; inhibits the activity of mitochondrial CS and GDH isolated from rat brain (IC50s = 437 and 420 µM, respectively); inhibits FadR from binding its promoter in E. coli (Ki = 2 µM); produced during isomerase-dependent β-oxidation of oleic acid in isolated rat heart mitochondria
Catalog No. 50-293-8528 Supplier Cayman Chemical Supplier No. 286075 MG
A thioester of decanoic acid and CoA; inhibits the activity of mitochondrial CS and GDH isolated from rat brain (IC50s = 437 and 420 µM, respectively); inhibits FadR from binding its promoter in E. coli (Ki = 2 µM); produced during isomerase-dependent β-oxidation of oleic acid in isolated rat heart mitochondria