Learn More
Cayman Chemical Decanoyl-L-carnitine-d3 (chloride), Purity: ≥99% deuterated forms (d1-d3), CAS: 2483831-87-2, 5 mg
SDP
Catalog No. 502935474
Encompass_Preferred
An internal standard for the quantification of decanoyl-L-carnitine by GC- or LC-MS
Catalog No. 50-293-5474 Supplier Cayman Chemical Supplier No. 265685 MG
An internal standard for the quantification of decanoyl-L-carnitine by GC- or LC-MS