Learn More
Cayman Chemical Decnoyl-L-carnItInchlorId 10mg
SDP
Catalog No. 502934549
Encompass_Preferred
A derivative of L-carnitine; increases the formation of C24 fatty acid intermediates, as well as docosapentaenoic and docosahexaenoic acid in rat hepatocytes
Catalog No. 50-293-4549 Supplier Cayman Chemical Supplier No. 2654910 MG
A derivative of L-carnitine; increases the formation of C24 fatty acid intermediates, as well as docosapentaenoic and docosahexaenoic acid in rat hepatocytes