Learn More
Cayman Chemical Decnoyl m-NItronIlIn 10mg
SDP
Catalog No. 502892908
Encompass_Preferred
One of several nitroaniline fatty acid amides that can be used to measure FAAH activity; exposure to FAAH results in the release of the yellow colorimetric dye m-nitroaniline (ε = 13,500 at 410 nm) allowing for fast and convenient measurement of FAAH activity using a 96-well plate spectrophotometer
Catalog No. 50-289-2908 Supplier Cayman Chemical Supplier No. 9034910 MG
One of several nitroaniline fatty acid amides that can be used to measure FAAH activity; exposure to FAAH results in the release of the yellow colorimetric dye m-nitroaniline (ε = 13,500 at 410 nm) allowing for fast and convenient measurement of FAAH activity using a 96-well plate spectrophotometer