Learn More
Cayman Chemical Decanoyl m-Nitroaniline, Purity: ≥98%, CAS: 72298-61-4, 50 mg
SDP
Catalog No. 502892911
Encompass_Preferred
One of several nitroaniline fatty acid amides that can be used to measure FAAH activity; exposure to FAAH results in the release of the yellow colorimetric dye m-nitroaniline (ε = 13,500 at 410 nm) allowing for fast and convenient measurement of FAAH activity using a 96-well plate spectrophotometer
Catalog No. 50-289-2911 Supplier Cayman Chemical Supplier No. 9034950 MG
One of several nitroaniline fatty acid amides that can be used to measure FAAH activity; exposure to FAAH results in the release of the yellow colorimetric dye m-nitroaniline (ε = 13,500 at 410 nm) allowing for fast and convenient measurement of FAAH activity using a 96-well plate spectrophotometer