Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Decoding Fragile X Syndrome
SDPSCI_ED

Catalog No. S001437 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S001437 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001437 Supplier Edvotek™ Supplier No. S55
Add to Cart
Add to Cart

In this kit, students gain an understanding of the screening process for Fragile X syndrome. Students will analyze simulated patient PCR samples to determine CGG repeat expansions in the FMR1 gene. Next, simulated Southern blot analysis will be used to confirm repeat size and methylation status.

  • Students will explore how CGG repeat expansions in the FMR1 gene cause Fragile X Syndrome, including the differences between normal, gray zone, premutation, and full mutation alleles
  • Gain hands-on experience preparing and running an agarose gel to separate simulated patient PCR samples, then interpret fluorescent band patterns to estimate CGG repeat counts and infer a patient's genetic sex
  • Learn how polymerase chain reaction amplifies specific regions of DNA to screen for Fragile X carrier status, and understand the limitations of PCR when it comes to detecting large, full-mutation expansions
  • Analyze simulated Southern blot results to detect large CGG repeat expansions and assess FMR1 gene methylation status
  • Group size: For 10 Lab Groups
  • Kit includes: Instructions, Ready-to-Load QuickStrip™ Dye Samples, UltraSpec-Agarose™, Electrophoresis Buffer (50X), Practice Gel Loading Solution, Disposable Pipets
  • All you need: DNA Electrophoresis Apparatus and Power Supply, Micropipettes: 5-50 μL (Optional), Blue Light Transilluminator, Microwave or Hot Plate
  • Storage: Room Temperature Stable. Storage of Ready-to-Load QuickStrip™ samples in the Refrigerator is Recommended

Specifications

Grade Level High School, Higher Ed
Class Size 10 Lab Groups
Product Type DNA Fingerprinting By Southern Blot
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.