Learn More
Werner Co. DeCoil Lanyard
Catalog No. 17103815
Change view
Click to view available options
Material:
Polyester Webbing
Catalog No. Material
17-103-815 Polyester Webbing
1 options
Catalog No. 17-103-815 Supplier Werner Co. Supplier No. C311104
  • $72.50
    $67.65 / Each of 1
    Save $4.85
    7% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
DeCoil lanyard with snap hook.
Werner DCELL Shock Pack is a deceleration device designed for controlled tearing action to reduce fall arrest forces on the user.
  • WebAlert inspectable polyester webbing makes wear and damage visible and obvious, making inspection easier, while providing durability that will last.
  • Snap hook is designed to prevent false engagement when connecting to the harness.
Specifications
Includes Snap Hook
Leg Configuration Single
Material Polyester Webbing
Connector Types Steel Snap Hook
Webbing Material Polyester
Color Gold, Blue
Length (English) 36 in.
Length (Metric) 91.44 cm
Certifications/Compliance ANSI Z359.13-2013, OSHA 29 CFR, 1910.140 / 1926.502
Product Type DeCoil Lanyard