Learn More
Werner Co. DeCoil Stretch Single Leg Lanyard
Description
Werner DCELL Shock Pack is a deceleration device designed for controlled tearing action to reduce fall arrest forces on the user.
- Elastic webbing contracts lanyard length from 6 to 4 ft. to reduce the hazards of tripping, dragging, and snagging
- Double Locking snap hook with 3,600 lb. gate strength
Specifications
Specifications
| Leg Configuration | Single |
| Load Bearing Capacity (Metric) | 1.633 kg Gate Strength |
| Length (Metric) | 182.88 cm |
| Product Type | DeCoil Lanyard |
| Length (English) | 72 in. |
| Connector Types | Snap Hook |
| Webbing Material | Elastic |
| Includes | Snap Hook |
| Load Bearing Capacity (English) | 3,600 lb Gate Strength |
| Color | Gold |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.