Learn More
LifeSensors PA950 Decomplexing Buffer 2mL
SDP
Catalog No. NC3372133
Encompass_Preferred
Decomplexing Buffer designed to be used with PA950. This component is designed for dilution and use after the cell lysate is generated to remove any native complexes that exist that cause high background. Please refer to the PA950 manual.
Catalog No. NC3372133 Supplier LifeSensors Supplier No. UE10032000
Decomplexing Buffer designed to be used with PA950. This component is designed for dilution and use after the cell lysate is generated to remove any native complexes that exist that cause high background. Please refer to the PA950 manual.