Learn More
Perkin Elmer US LLC DECON 90 Laboratory Detergent - Qty. 1 L
Catalog No. NC1666612
Encompass_Preferred
Decon 90 is the leading surface active cleaning agent, and/or radioactive decontaminant, for laboratory, medical and specialised industrial applications. It can be used for 'manual' cleaning or in an ultrasonic cleaning bath. It is biodegradable, phosphate-free, non-flammable and totally rinsable.
Catalog No. NC1666612 Supplier Perkin Elmer US LLC Supplier No. 09923462
Decon 90 is the leading surface active cleaning agent, and/or radioactive decontaminant, for laboratory, medical and specialised industrial applications. It can be used for 'manual' cleaning or in an ultrasonic cleaning bath. It is biodegradable, phosphate-free, non-flammable and totally rinsable.