Learn More
Electron Microscopy Sciences Decon 90 Cleaning Agent 1 LT
SDP
Supplier:  Electron Microscopy Sciences 6050501
Encompass_Preferred
Decon 90 is the leading surface active cleaning agent, and/or radioactive decontaminant, for laboratory, medical and specialised industrial applications. It can be used for ʽmanual’ cleaning or in an ultrasonic cleaning bath. It is biodegradable, phosphate-free, Bactericidal, non-flammable and totally rinsable.
Decon is an emulsion of anionic and non-ionic surface active agents, stabilising agents, non-phosphate detergent builders, alkalis and sequestering agents, in an aqueous base. Is NOT domestic applications. Suitable for glassware, ceramics, rubbers, plastics, stainless steel and ferrous metals. Being alkaline with pH in excess of pH 13, it is NOT suitable for use on non-ferrous metals or polycarbonate.
Catalog No. 50-337-35