Learn More
Dqe Supersoft Decon Brush
SDP
Catalog No. NC0791770
Encompass
Supersoft decon brushes are gentle on both protective clothing and skin. Available in 8" or 10" for use with either the Decon Brush Handle(HM544) or the Telescoping Decon Brush Handle(HM545). Available As Supersoft Decon Brush - 8" (HM540) Supersoft Decon Brush - 10" (HM541) Specifications Supersoft Decon Brush (HM540), (HM541) Durable to decontaminate HazMat suits, but gentle enough for decontaminating skin. Flow-thru capability (Flow-thru handle sold separately) Available in 8" or 10' Contents One brush head Additional Info public Available for International Sale
Catalog No. NC0791770 Supplier Dqe Supplier No. HM540
Supersoft decon brushes are gentle on both protective clothing and skin. Available in 8" or 10" for use with either the Decon Brush Handle(HM544) or the Telescoping Decon Brush Handle(HM545). Available As Supersoft Decon Brush - 8" (HM540) Supersoft Decon Brush - 10" (HM541) Specifications Supersoft Decon Brush (HM540), (HM541) Durable to decontaminate HazMat suits, but gentle enough for decontaminating skin. Flow-thru capability (Flow-thru handle sold separately) Available in 8" or 10' Contents One brush head Additional Info public Available for International Sale