Learn More
Dqe Decon Elevation Grid
SDP
Catalog No. NC0332778
Encompass
HazMat Elevation Grid Assemblies, made from sturdy, high-density polyethylene, are specifically designed for use with the DQE Standard Collection Pools and Compact Collection Pools. Wide and flat rubber feet protect pool liners and improve grid stability on uneven surfaces. Specifications Decon Elevation Grid (HM1023) Durable high-density polyethylene grid material Wide rubber feet inserts protect pool liners and tarp surfaces Flow-through design Dimensions: HM1023 - Two interlocked decking layers 3' x 2' x 5.5” Contents One grid assembly Additional Info AEL# 08D2-03-TDEW (Equipment, Technical Decontamination - Wet)
Catalog No. NC0332778 Supplier Dqe Supplier No. HM1023
HazMat Elevation Grid Assemblies, made from sturdy, high-density polyethylene, are specifically designed for use with the DQE Standard Collection Pools and Compact Collection Pools. Wide and flat rubber feet protect pool liners and improve grid stability on uneven surfaces. Specifications Decon Elevation Grid (HM1023) Durable high-density polyethylene grid material Wide rubber feet inserts protect pool liners and tarp surfaces Flow-through design Dimensions: HM1023 - Two interlocked decking layers 3' x 2' x 5.5” Contents One grid assembly Additional Info AEL# 08D2-03-TDEW (Equipment, Technical Decontamination - Wet)