Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decon™ Enzyte™ Enzyme Cleaner

Catalog No. 04355102 Shop All Decon Labs Products
Change view
Click to view available options
Volume (English):
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
04-355-102 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-355-102 Supplier Decon™ Supplier No. 1901

missing translation for 'validMainframeMsg'

Removes proteins, blood, and other organic materials from labware and hard surfaces without scrubbing

Ideal for soaking, ultrasonic cleaning, and automatic washers.

  • Low-foaming multi-enzyme cleaner
  • Concentrated—use 1 oz. per gallon of water
  • Completely rinsable
  • Noncaustic, nontoxic, and biodegradable
  • Unlimited shelf life
  • Environmentally Friendly: Item contains environmental attributes or features (e.g. water conserving, lead-free, etc.) as described in the item detail
  • Recycled: Item contains recycled content as described in the item detail
  • Non-toxic: Item is non-toxic

Specifications

Color Amber
Format Liquid
For Use With (Application) For removal of Proteins, Blood, fatty tissues
Volume (English) 1 gal.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.