Learn More
Thermo Scientific™ Decontamination Kit
GSA_VA
Catalog No. 15497025 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Decontamination Kit
Catalog No. Type
15-497-025 Decontamination Kit
1 options
This item is not returnable. View return policy
Catalog No. 15-497-025 Supplier Thermo Scientific™ Supplier No. 190651
Add to Cart
Add to Cart
This item is not returnable. View return policy
For Use With: For Water Jacket CO2 Incubators
For Use With: For Water Jacket CO2 Incubators
HEPA Filters
  • 99.97% filtering at 0.3μm
  • Compatible with all Thermo Scientific™ Forma™ Series II 3110, Thermo Scientific™ Steri-Cycle™, and Forma™ 310 Series (with HEPA filtration system option) incubators
  • Main chamber filter available as a stand-alone or packaged in a group with the other chamber in-line filters
Decontamination Kits
  • Consists of filters for difficult-to-clean chambers and gaskets (except door) and tubing
  • Reduces downtime and labor spent on decontamination
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description Internal chamber replacement kit with HEPA, gaskets, and other parts
For Use With (Equipment) For Water Jacket CO2 Incubators
Lighting LED
Type Decontamination Kit