Learn More
VAI DECON-PHENE PLUS Non-Sterile, 1 Gallon Concentrate
Catalog No. 19546432 Shop All VAI Products
Change view
Click to view available options
Volume (English):
1 gal.
Catalog No. Volume (English)
19-546-432 1 gal.
1 options
This item is not returnable. View return policy
Catalog No. 19-546-432 Supplier VAI Supplier No. DPP01
Add to Cart
Add to Cart
This item is not returnable. View return policy
A high pH phenolic that is compatible with most non-porous hard surface materials and can be used on surfaces in aseptic filling and gowning rooms, in general manufacturing areas, and in laboratories.
  • Effective sanitizer on inanimate, non-porous, non-food contact surfaces
  • Sterile pharmaceutical cleanroom formula
  • Phosphate free
Specifications
Product Type Phenolic Disinfectant
Form Clear Liquid
Scent Unscented
Sterility Non-sterile
Volume (English) 1 gal.
WARNING: Cancer - www.P65Warnings.ca.gov