Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAI DECON-PHENE PLUS Non-Sterile with Attached Trigger, 16 oz. SimpleMix

Catalog No. 19546443 Shop All VAI Products
Change view
Click to view available options
Dilution:
1:128
1:64
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dilution
19-546-443 1:64
19-546-442 1:128
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-546-443 Supplier VAI Supplier No. DPP0716Z02
Add to Cart
Add to Cart

A high pH phenolic that is compatible with most non-porous hard surface materials and can be used on surfaces in aseptic filling and gowning rooms, in general manufacturing areas, and in laboratories.

  • Effective sanitizer on inanimate, non-porous, non-food contact surfaces
  • Sterile pharmaceutical cleanroom formula
  • Phosphate free

Specifications

Product Type Phenolic Disinfectant
Form Clear Liquid
Scent Unscented
Dilution 1:64
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.