Learn More
Grainger Florock Decorative Quartz Aggregate, Slip Resistant, Gray, 50 lb.
Catalog No. 502783454
Encompass_Preferred
This decorative, aggregate-filled system provides a highly durable, seamless, and sanitary flooring solution. Engineered for strength and wear resistance, it delivers a slip-resistant and impact-resistant surface suitable for a wide range of institutional, commercial, and industrial settings, including laboratories and food processing areas.
  • Provides a slip-resistant and impact-resistant surface
  • Features exceptional strength and wear characteristics
  • Offers a seamless, sanitary finish
  • Chemical-resistant properties
  • Suitable for use in heavy traffic areas
  • Complies with ADA, EPA, FDA, OSHA, and USDA standards
  • Versatile application for interior and exterior environments
Catalog No. 50-278-3454 Supplier Grainger Supplier No. 19NZ49
This decorative, aggregate-filled system provides a highly durable, seamless, and sanitary flooring solution. Engineered for strength and wear resistance, it delivers a slip-resistant and impact-resistant surface suitable for a wide range of institutional, commercial, and industrial settings, including laboratories and food processing areas.
  • Provides a slip-resistant and impact-resistant surface
  • Features exceptional strength and wear characteristics
  • Offers a seamless, sanitary finish
  • Chemical-resistant properties
  • Suitable for use in heavy traffic areas
  • Complies with ADA, EPA, FDA, OSHA, and USDA standards
  • Versatile application for interior and exterior environments