Learn More
Grainger Florock Decorative Quartz, Beige, Silica, 50 lb.
Catalog No. 502782671
Encompass_Preferred
These decorative quartz floor chips are designed to add color and texture to resin-coated flooring while increasing the surface's durability and impact resistance. Ideal for both interior and exterior use, the aggregate is suitable for heavy traffic areas.
  • Provides impact and slip resistance
  • Resistant to chemicals and mildew
  • Suitable for interior and exterior environments
  • Requires a primer prior to application
  • Application temperature range of 55°F to 95°F
  • Features a dry time of 6 to 24 hr
  • Meets ADA, EPA, FDA, OSHA, and USDA standards
Catalog No. 50-278-2671 Supplier Grainger Supplier No. 19NZ46
These decorative quartz floor chips are designed to add color and texture to resin-coated flooring while increasing the surface's durability and impact resistance. Ideal for both interior and exterior use, the aggregate is suitable for heavy traffic areas.
  • Provides impact and slip resistance
  • Resistant to chemicals and mildew
  • Suitable for interior and exterior environments
  • Requires a primer prior to application
  • Application temperature range of 55°F to 95°F
  • Features a dry time of 6 to 24 hr
  • Meets ADA, EPA, FDA, OSHA, and USDA standards