Learn More
Grainger Florock Decorative Quartz Aggregate, Silica, Tan, 50 lb
Catalog No. 502783456
Encompass_Preferred
These decorative silica quartz chips are designed to enhance resin-coated flooring systems by providing vibrant color, improved texture, and increased durability. Suitable for both interior and exterior applications, this aggregate withstands heavy traffic and offers chemical and mildew resistance.
  • Enhances floor durability and impact resistance
  • Suitable for heavy traffic areas
  • Provides chemical and mildew resistance
  • Designed for broadcast application
  • Covers 100 sq. ft.
  • Cleaning via vacuum
Catalog No. 50-278-3456 Supplier Grainger Supplier No. 19NZ50
These decorative silica quartz chips are designed to enhance resin-coated flooring systems by providing vibrant color, improved texture, and increased durability. Suitable for both interior and exterior applications, this aggregate withstands heavy traffic and offers chemical and mildew resistance.
  • Enhances floor durability and impact resistance
  • Suitable for heavy traffic areas
  • Provides chemical and mildew resistance
  • Designed for broadcast application
  • Covers 100 sq. ft.
  • Cleaning via vacuum