Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Decorin (DCN) ELISA Kit

Catalog No. EHDCNX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EHDCNX5 5 x 96 Tests
EHDCN 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EHDCNX5 Supplier Invitrogen™ Supplier No. EHDCNX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

ELISA

The Human Decorin (Hu DCN) ELISA quantitates Hu DCN in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu DCN. Principle of the method The Human DCN solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Decorin is a member of the small, leucine-rich proteoglycan (SLRP) family along with biglycan. Found in the interterritorial region of cartilage, decorin interacts with collagen and other matrix proteins to regulate the cartilaginous matrix. It has been shown to interact with fibronectin, thrombospondin, C1q, epidermal growth factor receptor (EGFR), and TGF-b. It is believed that decorin also plays a role in the cell cycle of chondrocytes. The decorin neoepitope fragment has a sequence of PKTLQE, and is generated by MMP-3 or ADAMTS-4 cleavage at Glu154-Leu155 of decorin core protein.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P07585
Assay Range 1.5-700 pg/mL
Assay Sensitivity 1.5 pg/mL
Calibration Range 1.5 pg/mL to 700 pg/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Cell Structure/Organelle Proteins
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA kit shows no cross-reactivity with any of the following cytokines tested: human Angiogenin, BDNF, BLC, ENA-78, FGF-4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-10, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, G-CSF, GM-CSF, IFN-gamma, Leptin (OB), MCP-1, MCP-3, MDC, MIP-1 alpha, MIP-1 beta, MIP-1 delta, MMP-1, -2, -3, -10, PARC, RANTES, SCF, TARC, TGF-beta, TIMP-1, TIMP- 2, TNF-alpha, TNF-beta, TPO, VEGF
Sufficient For 5 x 96 assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias Bone proteoglycan II; CSCD; DC; Dcn; DCN protein; DCN1C; Decorin; decorin isoform a preproprotein; Dermatan sulfate proteoglycan-II; dermatan sulfate proteoglycan-II (decorin); dermatan sulphate proteoglycans II; Dermatan sulphate proteoglycans II (DSPG2); DSPG; DSPG2; mDcn; PG40; PGII; PGS2; PG-S2; proteoglycan core protein; proteoglycan core protein; binds type I collagen fibers and TGF-beta; decoron; similar to bone proteoglycan II; similar to decorin; SLRR1B; Small leucine rich protein 1B; small leucine-rich protein 1B; unnamed protein productBone proteoglycan II; CSCD; DC; Dcn; DCN protein; DCN1C; Decorin; decorin isoform a preproprotein; Dermatan sulfate proteoglycan-II; dermatan sulfate proteoglycan-II (decorin); dermatan sulphate proteoglycans II; Dermatan sulphate proteoglycans II (DSPG2); DSPG; DSPG2; mDcn; PG40; PGII; PGS2; PG-S2; proteoglycan core protein; proteoglycan core protein; binds type I collagen fibers and TGF-beta; decoron; similar to bone proteoglycan II; similar to decorin; SLRR1B; Small leucine rich protein 1B; small leucine-rich protein 1B; unnamed protein product; CSCD; DSPG2; PG40; PGII; PGS2; SLRR1B
Gene ID (Entrez) 1634
Gene Symbol DCN
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases Bone proteoglycan II, PG-S2, PG40
Quantity 5 x 96 Tests
Regulatory Status RUO
Research Discipline Extracellular Matrix & Cell Adhesion
Sample Volume Plasma 2 μL, Serum 2 μL, Supernatant 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Synonym CSCD, DSPG2, PG40, PGII, PGS2, SLRR1B
Target Decorin
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.