Learn More
Sigma Aldrich Fine Chemicals Biosciences Decorin from bovine articu5MG
Catalog No. 502905196
Encompass
Decorin from bovine articular cartilage is a proteoglycan that belongs to the small leucine-rich repeat proteoglycans and proteins (SLRPs) family. Decorin is present in almost all tissues. It has a protein core composed of leucine-rich repeats (LRRs) flanked by two cysteine-rich regions.
Catalog No. 50-290-5196 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D8428.5MG
Decorin from bovine articular cartilage is a proteoglycan that belongs to the small leucine-rich repeat proteoglycans and proteins (SLRPs) family. Decorin is present in almost all tissues. It has a protein core composed of leucine-rich repeats (LRRs) flanked by two cysteine-rich regions.