Learn More
Cayman Chemical DecorIn Long Isoformhumn100ug
SDP
Catalog No. 502946165
Encompass_Preferred
Source: Recombinant human C-terminal His-tagged decorin expressed in HEK293 cells • Amino acids: 17-359 • MW: 39.4 kDa
Catalog No. 50-294-6165 Supplier Cayman Chemical Supplier No. 32095100 UG
Source: Recombinant human C-terminal His-tagged decorin expressed in HEK293 cells • Amino acids: 17-359 • MW: 39.4 kDa