Learn More
Cayman Chemical Decorin Long Isoform (human, recombinant), Purity: ≥95% estimated by SDS-PAGE, 1 mg
SDP
Catalog No. 502950578
Encompass_Preferred
Source: Recombinant human C-terminal His-tagged decorin expressed in HEK293 cells • Amino acids: 17-359 • MW: 39.4 kDa
Catalog No. 50-295-0578 Supplier Cayman Chemical Supplier No. 320951 MG
Source: Recombinant human C-terminal His-tagged decorin expressed in HEK293 cells • Amino acids: 17-359 • MW: 39.4 kDa