Learn More
ABclonal Technology Decorin Rabbit mAb
SDP
Catalog No. 5000008415
Encompass_Preferred
This gene encodes a member of the small leucine-rich proteoglycan family of proteins Alternative splicing results in multiple transcript variants at least one of which encodes a preproprotein that is proteolytically processed to generate the mature protein This protein plays a role in collagen fibril assembly Binding of this protein to multiple cell surface receptors mediates its role in tumor suppression including a stimulatory effect on autophagy and inflammation and an inhibitory effect on angiogenesis and tumorigenesis This gene and the related gene biglycan are thought to be the result of a gene duplication Mutations in this gene are associated with congenital stromal corneal dystrophy in human patients
Catalog No. 50-000-08415 Supplier ABclonal Technology Supplier No. A26852100UL
This gene encodes a member of the small leucine-rich proteoglycan family of proteins Alternative splicing results in multiple transcript variants at least one of which encodes a preproprotein that is proteolytically processed to generate the mature protein This protein plays a role in collagen fibril assembly Binding of this protein to multiple cell surface receptors mediates its role in tumor suppression including a stimulatory effect on autophagy and inflammation and an inhibitory effect on angiogenesis and tumorigenesis This gene and the related gene biglycan are thought to be the result of a gene duplication Mutations in this gene are associated with congenital stromal corneal dystrophy in human patients