Learn More
Medchemexpress LLC HY-101835 1mg Medchemexpress, Decoyinine CAS:2004-04-8 Purity:>98%
SDP
Catalog No. 501967881
Encompass_Preferred
Medchemexpress, HY-101835 1mg Decoyinine CAS:2004-04-8 Decoyinine is a selective inhibitor of GMP synthetase (GMPS). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7881 Supplier Medchemexpress LLC Supplier No. HY1018351MG
Medchemexpress, HY-101835 1mg Decoyinine CAS:2004-04-8 Decoyinine is a selective inhibitor of GMP synthetase (GMPS). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.