Learn More
AdipoGen Decoyinine Angustmycin A
SDP
Supplier:  AdipoGen BVT0030M001
Encompass_Preferred
Chemical. CAS 2004-04-8. Formula C11H13N5O4. MW 279.3. Isolated from Streptomyces sp. S 2113. Nucleoside antibiotic. Antitumor compound. Xanthosine monophosphate XMP aminase inhibitor. RNA synthesis inhibitor. Specific GMP synthase inhibitor. Reduces intracellular GTP levels.
Catalog No. 50-256-2137