Learn More
AdipoGen Decoyinine Angustmycin A
SDP
Catalog No. 502562136
Encompass_Preferred
Chemical. CAS 2004-04-8. Formula C11H13N5O4. MW 279.3. Isolated from Streptomyces sp. S 2113. Nucleoside antibiotic. Antitumor compound. Xanthosine monophosphate XMP aminase inhibitor. RNA synthesis inhibitor. Specific GMP synthase inhibitor. Reduces intracellular GTP levels.
Catalog No. 50-256-2136 Supplier AdipoGen Supplier No. BVT0030M005
Chemical. CAS 2004-04-8. Formula C11H13N5O4. MW 279.3. Isolated from Streptomyces sp. S 2113. Nucleoside antibiotic. Antitumor compound. Xanthosine monophosphate XMP aminase inhibitor. RNA synthesis inhibitor. Specific GMP synthase inhibitor. Reduces intracellular GTP levels.