Learn More
Aviva Systems Biology Corp Decr2 Antibody - C-terminal region (ARP43567_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43567_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Decr2. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: -ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rat, Yeast
Catalog No. 50-285-082