Learn More
Electron Microscopy Sciences Serum Vial De-Crimpier Pliers for sizes 8 and 13 mm
SDP
Catalog No. 5032784
Encompass_Preferred
Made from stainless steel offers one tool for both sizes: 13 mm and 20 mm serum vial de-crimpier. An inexpensive and easy way to remove the seal from the serum vial, simply secure the seal between the jaws, squeeze the handle and twist the seal. Plastic coated handle offers comfort and ease of use.
Use: 8 and 13 mm
Catalog No. 50-327-84 Supplier Electron Microscopy Sciences Supplier No. 6218001
Made from stainless steel offers one tool for both sizes: 13 mm and 20 mm serum vial de-crimpier. An inexpensive and easy way to remove the seal from the serum vial, simply secure the seal between the jaws, squeeze the handle and twist the seal. Plastic coated handle offers comfort and ease of use.
Use: 8 and 13 mm