Learn More
Cliawaived Inc Dectabuse Fentanyl 5ng/mL 5mL Positive Control Solution for 1ng/mL Fentanyl rapid test kits
SDP
Catalog No. NC2684683
Encompass_Preferred
Biochemical Diagnostics DETECTABUSE liquid urine controls are designed to monitor the performance of different quantitative or qualitative urinary drug detection methods such as GC/MS, LC/MS, HPLC and/or Immunoassay screening methods. Our human urine matrix mimics patient samples for an accurate external third-party quality control formulated with various drugs to meet the testing requirements of most testing labs. Note: Items only ships Next Day Air and Refrigerated.
Catalog No. NC2684683 Supplier Cliawaived Inc Supplier No. BIOC18008613KIT
Biochemical Diagnostics DETECTABUSE liquid urine controls are designed to monitor the performance of different quantitative or qualitative urinary drug detection methods such as GC/MS, LC/MS, HPLC and/or Immunoassay screening methods. Our human urine matrix mimics patient samples for an accurate external third-party quality control formulated with various drugs to meet the testing requirements of most testing labs. Note: Items only ships Next Day Air and Refrigerated.