Learn More
TARGETMOL CHEMICALS INC DECURSIN 10MG
SDP
Catalog No. 502576837
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Decursin (Decursinol angelate) is able to attenuate kainic acid-induced seizures and could have potential as an antiepileptic drug. 2. Decursin exhibits hepatoprotective effects potentially by inhibiting the TGF-(beta)1 induced NOX activation and Smad signaling. 3. Decursin has anti-cancer activity mediated suppression of the PKC(alpha) MAPK and NF-(kappa)B pathways in MCF-7 cells. 4. Decursin exhibits cytotoxicity against various human cancer cells and to possess anti-amnesic activity in vivo through the inhibition of AChE activity. purity: 97%
Catalog No. 50-257-6837 Supplier TARGETMOL CHEMICALS INC Supplier No. T3S141610MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Decursin (Decursinol angelate) is able to attenuate kainic acid-induced seizures and could have potential as an antiepileptic drug. 2. Decursin exhibits hepatoprotective effects potentially by inhibiting the TGF-(beta)1 induced NOX activation and Smad signaling. 3. Decursin has anti-cancer activity mediated suppression of the PKC(alpha) MAPK and NF-(kappa)B pathways in MCF-7 cells. 4. Decursin exhibits cytotoxicity against various human cancer cells and to possess anti-amnesic activity in vivo through the inhibition of AChE activity. purity: 97%