Learn More
Medchemexpress LLC (±)-Decursinol | 5993-18-0 | 5 MG
SDP
Catalog No. 5000420316
Encompass_Preferred
(±)-Decursinol is a potent FtsZ inhibitor. It inhibits *B. anthracis* FtsZ polymerization with an IC50 of 102 μM. This compound is provided as a white to light yellow solid.
  • Potent FtsZ inhibitor
  • Inhibits *B. anthracis* FtsZ polymerization with an IC50 of 102 μM
  • White to light yellow solid
  • For research use only
Catalog No. 50-004-20316 Supplier Medchemexpress LLC Supplier No. HYN25675MG
(±)-Decursinol is a potent FtsZ inhibitor. It inhibits *B. anthracis* FtsZ polymerization with an IC50 of 102 μM. This compound is provided as a white to light yellow solid.
  • Potent FtsZ inhibitor
  • Inhibits *B. anthracis* FtsZ polymerization with an IC50 of 102 μM
  • White to light yellow solid
  • For research use only