Learn More
Medchemexpress LLC HY-N4109 5mg Medchemexpress, Decursinol CAS:23458-02-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN41095MG
Encompass_Preferred
Medchemexpress, HY-N4109 5mg Decursinol CAS:23458-02-8 Decursinol, isolated from the roots of Angelica gigas, possesses antinociceptive effect with orally bioavailability. Decursinol possesses anti-tumor and anti-metastasis activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0421