Learn More
Medchemexpress LLC Decursinol angelate | 130848-06-5 | MFCD06796687 | 99.9% | 328.36 g/mol | C19H20O5 | 10 MG
SDP
Catalog No. 5000289157
Encompass_Preferred
Decursinol angelate is a natural coumarin derivative isolated from the root of Angelica gigas. It functions as a protein kinase C (PKC) activator and has reported cytotoxic, anti-tumor, and anti-inflammatory activities. The compound is supplied as a white to off-white solid and is used in biochemical and cellular research.
  • Acts as a protein kinase C (PKC) activator.
  • Reported cytotoxic and anti-tumor activity in published studies.
  • High reported purity suitable for analytical and biological assays.
  • White to off-white solid with recommended storage at 4°C, protected from light.
  • Available in small-mass sizes for laboratory experiments.
Catalog No. 50-002-89157 Supplier Medchemexpress LLC Supplier No. HYN432210MG
Decursinol angelate is a natural coumarin derivative isolated from the root of Angelica gigas. It functions as a protein kinase C (PKC) activator and has reported cytotoxic, anti-tumor, and anti-inflammatory activities. The compound is supplied as a white to off-white solid and is used in biochemical and cellular research.
  • Acts as a protein kinase C (PKC) activator.
  • Reported cytotoxic and anti-tumor activity in published studies.
  • High reported purity suitable for analytical and biological assays.
  • White to off-white solid with recommended storage at 4°C, protected from light.
  • Available in small-mass sizes for laboratory experiments.