Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decyl alcohol, 98+%

Catalog No. p-3736091
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC165760010 1 L Glass Bottle
AC165760025 2.5 L Glass Bottle
AC165760100 10 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC165760010 Supplier Thermo Scientific Chemicals Supplier No. 165760010
Only null left
Add to Cart

CAS: 112-30-1 | C10H22O | 158.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-30-1
Molecular Formula C10H22O
Molecular Weight (g/mol) 158.28
MDL Number MFCD00004747
InChI Key MWKFXSUHUHTGQN-UHFFFAOYSA-N
Synonym 1-decanol, decyl alcohol, decanol, n-decyl alcohol, n-decanol, capric alcohol, nonylcarbinol, antak, caprinic alcohol, royaltac
PubChem CID 8174
ChEBI CHEBI:28903
IUPAC Name decan-1-ol
SMILES CCCCCCCCCCO

Specifications

Melting Point 6°C
Color Colorless to Yellow
Density 0.8290g/mL
Boiling Point 220°C to 240°C
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4360 to 1.438
Linear Formula CH3(CH2)9OH
Quantity 1 L
Beilstein 01, 425
Specific Gravity 0.829
Merck Index 15, 2858
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone,benzene,chlorogorm,soluble in alcohol,ether,mineral oil,propylene,insoluble in glycerin water (233°C)
Viscosity 14.1 mPa.s (20°C)
Formula Weight 158.28
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Decyl alcohol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protec
EINECSNumber 203-956-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.