Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decyl Alcohol 99+%, Thermo Scientific™

Catalog No. AC219630500
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC219630500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC219630500 Supplier Thermo Scientific Chemicals Supplier No. 219630500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-30-1
Molecular Formula C10H22O
Molecular Weight (g/mol) 158.285
InChI Key MWKFXSUHUHTGQN-UHFFFAOYSA-N
Synonym 1-decanol, decyl alcohol, decanol, n-decyl alcohol, n-decanol, capric alcohol, nonylcarbinol, antak, caprinic alcohol, royaltac
PubChem CID 8174
ChEBI CHEBI:28903
IUPAC Name decan-1-ol
SMILES CCCCCCCCCCO

Specifications

Color Colorless to Yellow
Infrared Spectrum Authentic
Refractive Index 1.4360 to 1.4380 (20°C, 589nm)
Linear Formula CH3(CH2)9OH
Quantity 50 g
Beilstein 01, 425
Merck Index 15, 2858
Formula Weight 158.28
Percent Purity ≥99.0%
Physical Form Liquid
Chemical Name or Material Decyl alcohol, 99+%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS P Statement Causes serious eye irritation. Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement Avoid release to the environment. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clot
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.