Learn More
Medchemexpress LLC Decyl aldehyde | 112-31-2 | 99% | 156.269 | 1 G
SDP
Catalog No. 5000452805
Encompass_Preferred
Decyl aldehyde is a simple ten-carbon aldehyde and a bacterial luciferase substrate. It appears as a colorless to light yellow liquid with a density of 0.83 g/cm3. It is used for research purposes.
  • Simple ten-carbon aldehyde
  • Bacterial luciferase substrate
  • Appears as a liquid (density: 0.83 g/cm3)
  • Colorless to light yellow
  • For research use only
Catalog No. 50-004-52805 Supplier Medchemexpress LLC Supplier No. HYW0125701G
Decyl aldehyde is a simple ten-carbon aldehyde and a bacterial luciferase substrate. It appears as a colorless to light yellow liquid with a density of 0.83 g/cm3. It is used for research purposes.
  • Simple ten-carbon aldehyde
  • Bacterial luciferase substrate
  • Appears as a liquid (density: 0.83 g/cm3)
  • Colorless to light yellow
  • For research use only