Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Decyl aldehyde | 112-31-2 | 99% | 156.27 | 5 G
SDP

Catalog No. 5000453027
Encompass_Preferred

Decyl aldehyde is a simple ten-carbon aldehyde and a bacterial luciferase substrate. It is a colorless to light yellow liquid with a purity of 99%, commonly supplied as an endogenous metabolite.

  • A simple ten-carbon aldehyde
  • Functions as a bacterial luciferase substrate
  • Purity of 99%
  • Colorless to light yellow liquid appearance
  • Molecular formula C10H20O
  • Recommended storage at 4°C, stored under nitrogen
  • Soluble in DMSO at 100 mg/mL
  • Demonstrates biological activity against yeast cells and C. elegans

Catalog No. 50-004-53027 Supplier Medchemexpress LLC Supplier No. HYW0125705G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Decyl aldehyde is a simple ten-carbon aldehyde and a bacterial luciferase substrate. It is a colorless to light yellow liquid with a purity of 99%, commonly supplied as an endogenous metabolite.

  • A simple ten-carbon aldehyde
  • Functions as a bacterial luciferase substrate
  • Purity of 99%
  • Colorless to light yellow liquid appearance
  • Molecular formula C10H20O
  • Recommended storage at 4°C, stored under nitrogen
  • Soluble in DMSO at 100 mg/mL
  • Demonstrates biological activity against yeast cells and C. elegans

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.