Learn More
Medchemexpress LLC Decyl aldehyde | 112-31-2 | 99% | 156.27 | 500 MG
SDP
Catalog No. 5000455113
Encompass_Preferred
Decyl aldehyde is a simple ten-carbon aldehyde and a bacterial luciferase substrate.
  • Endogenous metabolite
  • Prevents growth of yeast cells and results in yeast cell mortality (at 5 μL, 0-500 min).
  • Toxicity to C. elegans (EC50 < 0.03% at 0-0.05%).
Catalog No. 50-004-55113 Supplier Medchemexpress LLC Supplier No. HYW012570500MG
Decyl aldehyde is a simple ten-carbon aldehyde and a bacterial luciferase substrate.
  • Endogenous metabolite
  • Prevents growth of yeast cells and results in yeast cell mortality (at 5 μL, 0-500 min).
  • Toxicity to C. elegans (EC50 < 0.03% at 0-0.05%).