Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Decyl β-D-maltopyranoside | 82494-09-5 | 482.56 g/mol | C22H42O11 | 100 MG
SDP

Catalog No. 5000208773
Encompass_Preferred

Decyl β-D-maltopyranoside is a nonionic biochemical reagent commonly used as a mild detergent to solubilize and stabilize membrane proteins in aqueous systems. It is supplied as a dry solid for laboratory research applications.

  • Nonionic detergent for membrane protein solubilization and stabilization
  • Soluble in aqueous buffers
  • Molecular formula C22H42O11 and molecular weight 482.56 g/mol
  • Available in a 100 mg package

Catalog No. 50-002-08773 Supplier Medchemexpress LLC Supplier No. HYW040203100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Decyl β-D-maltopyranoside is a nonionic biochemical reagent commonly used as a mild detergent to solubilize and stabilize membrane proteins in aqueous systems. It is supplied as a dry solid for laboratory research applications.

  • Nonionic detergent for membrane protein solubilization and stabilization
  • Soluble in aqueous buffers
  • Molecular formula C22H42O11 and molecular weight 482.56 g/mol
  • Available in a 100 mg package

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.