Learn More
Medchemexpress LLC HY-138887 25mg , Decyl maltose neopentyl glycol CAS:1257852-99-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13888725MG
Encompass_Preferred
HY-138887 25mg Decyl maltose neopentyl glycol CAS: 1257852-99-5 Decyl maltose neopentyl glycol (DMNG) is the neopentyl glycol detergent that does not disrupt the AlkB oligomeric state. AlkB is a nonheme di-iron alkane hydroxylase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1814