Learn More
Medchemexpress LLC Decyl Maltose Neopentyl Glycol (DMNG) | 1257852-99-5 | 98.0% | 949.08 | 1 ML
SDP
Catalog No. 5000352109
Encompass_Preferred
Decyl maltose neopentyl glycol (DMNG) is a neopentyl glycol detergent that does not disrupt the AlkB oligomeric state. AlkB is a nonheme di-iron alkane hydroxylase. It is for research use only.
  • Neopentyl glycol detergent
  • Does not disrupt AlkB oligomeric state
  • For research use only
Catalog No. 50-003-52109 Supplier Medchemexpress LLC Supplier No. HY13888710MM/1ML
Decyl maltose neopentyl glycol (DMNG) is a neopentyl glycol detergent that does not disrupt the AlkB oligomeric state. AlkB is a nonheme di-iron alkane hydroxylase. It is for research use only.
  • Neopentyl glycol detergent
  • Does not disrupt AlkB oligomeric state
  • For research use only