Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Decyl maltose neopentyl glycol (DMNG) | 1257852-99-5 | 98.0% | 949.08 | 50 MG
SDP

Catalog No. 5000350541
Encompass_Preferred

Decyl maltose neopentyl glycol (DMNG) is a neopentyl glycol detergent designed to preserve the oligomeric state and hydroxylase activity of AlkB, a nonheme di-iron alkane hydroxylase. It is suitable for research applications.

  • Preserves AlkB oligomeric state
  • Maintains P. putida AlkB hydroxylase activity after purification
  • Purity: 98.0%
  • Molecular weight: 949.08
  • Appearance: White to off-white solid

Catalog No. 50-003-50541 Supplier Medchemexpress LLC Supplier No. HY13888750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Decyl maltose neopentyl glycol (DMNG) is a neopentyl glycol detergent designed to preserve the oligomeric state and hydroxylase activity of AlkB, a nonheme di-iron alkane hydroxylase. It is suitable for research applications.

  • Preserves AlkB oligomeric state
  • Maintains P. putida AlkB hydroxylase activity after purification
  • Purity: 98.0%
  • Molecular weight: 949.08
  • Appearance: White to off-white solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.