Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Decyl methyl ether | 7289-52-3 | MFCD00216758 | >95.0% | 172.31 g/mol | C11H24O | 100 MG
SDP

Catalog No. 5000280469
Encompass_Preferred

Decyl methyl ether (1-methoxydecane) is a fatty ether used as a laboratory reagent, solvent, and fragrance ingredient reference. It is supplied in small laboratory packs; consult the certificate of analysis and SDS for lot-specific purity and handling.

  • Used as a solvent and biochemical assay reagent.
  • Synonym: 1-methoxydecane.
  • Chemical formula: C11H24O.
  • Molecular weight: 172.31 g/mol.
  • Reported purity: >95.0% by GC; consult CoA for lot values.
  • Available in small packs suitable for analytical and research use.

Catalog No. 50-002-80469 Supplier Medchemexpress LLC Supplier No. HYW587892100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Decyl methyl ether (1-methoxydecane) is a fatty ether used as a laboratory reagent, solvent, and fragrance ingredient reference. It is supplied in small laboratory packs; consult the certificate of analysis and SDS for lot-specific purity and handling.

  • Used as a solvent and biochemical assay reagent.
  • Synonym: 1-methoxydecane.
  • Chemical formula: C11H24O.
  • Molecular weight: 172.31 g/mol.
  • Reported purity: >95.0% by GC; consult CoA for lot values.
  • Available in small packs suitable for analytical and research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.