Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Decyl methyl ether | 7289-52-3 | MFCD00216758 | ≥98.0% | 172.31 | C11H24O | 25 MG
SDP

Catalog No. 5000285064
Encompass_Preferred

Decyl methyl ether (1-methoxydecane) is a fatty ether research reagent used in biochemical and fragrance-related applications. The compound is described as a strong fatty ether and is reported to be non-genotoxic and phototoxic/photosensitizing. It is supplied for laboratory research use only and should be handled according to safety data sheet recommendations.

  • Fatty ether reagent for research use only.
  • Chemical formula C11H24O; molecular weight 172.31.
  • CAS number 7289-52-3 for precise identification.
  • Available in small milligram and gram package sizes suitable for assay development.
  • Reported non-genotoxic; may be phototoxic/photosensitizing - handle with appropriate precautions.

Catalog No. 50-002-85064 Supplier Medchemexpress LLC Supplier No. HYW58789225MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Decyl methyl ether (1-methoxydecane) is a fatty ether research reagent used in biochemical and fragrance-related applications. The compound is described as a strong fatty ether and is reported to be non-genotoxic and phototoxic/photosensitizing. It is supplied for laboratory research use only and should be handled according to safety data sheet recommendations.

  • Fatty ether reagent for research use only.
  • Chemical formula C11H24O; molecular weight 172.31.
  • CAS number 7289-52-3 for precise identification.
  • Available in small milligram and gram package sizes suitable for assay development.
  • Reported non-genotoxic; may be phototoxic/photosensitizing - handle with appropriate precautions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.